Anti-ATP5B Rabbit Polyclonal Antibody

Toimittaja: Abgent
AP2973C
ABGEAP2973CEA 354 EUR
ABGEAP2973C
Anti-ATP5B Rabbit Polyclonal Antibody
Type: Primary
Antigen: ATP5B (ATP synthase, H+ transporting, mitochondrial F1 complex, beta polypeptide)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR