Anti-MGAT3 Rabbit Polyclonal Antibody

Toimittaja: Abgent
AP2411A
ABGEAP2411AEA 354 EUR
ABGEAP2411A
Anti-MGAT3 Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: MGAT3 (mannosyl (beta-1,4-)-glycoprotein beta-1,4-N-acetylglucosaminyltransferase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR