Anti-CTAGE1 Rabbit Polyclonal Antibody

Toimittaja: Abgent
AP20180C
ABGEAP20180CEA 354 EUR
ABGEAP20180C
Anti-CTAGE1 Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: CTAGE1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR