Anti-KIT Rabbit Polyclonal Antibody

Toimittaja: Abgent
AP19037B
ABGEAP19037BEA 354 EUR
ABGEAP19037B
Anti-KIT Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: KIT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR