Anti-KRT31 Rabbit Polyclonal Antibody

Toimittaja: Abgent
AP18844A
ABGEAP18844AEA 354 EUR
ABGEAP18844A
Anti-KRT31 Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: KRT31
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR