Anti-ZNRF4 Rabbit Polyclonal Antibody

Toimittaja: Abgent
AP18372A
ABGEAP18372AEA 354 EUR
ABGEAP18372A
Anti-ZNRF4 Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: ZNRF4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR