Anti-ZNF695 Rabbit Polyclonal Antibody

Toimittaja: Abgent
AP17947C
ABGEAP17947CEA 354 EUR
ABGEAP17947C
Anti-ZNF695 Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: ZNF695
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR