Anti-GRM6 Rabbit Polyclonal Antibody

Toimittaja: Abgent
AP1642A
ABGEAP1642AEA 354 EUR
ABGEAP1642A
Anti-GRM6 Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: GRM6 (glutamate receptor, metabotropic 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR