Anti-HRH2 Rabbit Polyclonal Antibody

Toimittaja: Abgent
AP14970C
ABGEAP14970CEA 354 EUR
ABGEAP14970C
Anti-HRH2 Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: HRH2 (histamine receptor H2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR