Anti-TPX2 Rabbit Polyclonal Antibody

Toimittaja: Abgent
AP14553C
ABGEAP14553CEA 354 EUR
ABGEAP14553C
Anti-TPX2 Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: TPX2 (TPX2, microtubule-associated)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR