Anti-RPA1 Rabbit Polyclonal Antibody
Toimittaja: Abgent
AP14415B
ABGEAP14415BEA 354 EUR
ABGEAP14415B
Anti-RPA1 Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: RPA1 (replication protein A1, 70kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human