Anti-SLC11A2 Rabbit Polyclonal Antibody

Toimittaja: Abgent
AP13801C
ABGEAP13801CEA 354 EUR
ABGEAP13801C
Anti-SLC11A2 Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: SLC11A2 (solute carrier family 11 (proton-coupled divalent metal ion transporter), member 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR