Anti-APOO Rabbit Polyclonal Antibody

Toimittaja: Abgent
AP13324A
ABGEAP13324AEA 354 EUR
ABGEAP13324A
Anti-APOO Rabbit Polyclonal Antibody
Type: Primary
Antigen: APOO (apolipoprotein O)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR