Anti-TBX6 Rabbit Polyclonal Antibody

Toimittaja: Abgent
AP11956B
ABGEAP11956BEA 354 EUR
ABGEAP11956B
Anti-TBX6 Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: TBX6 (T-box 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR