Anti-TTLL13 Rabbit Polyclonal Antibody

Toimittaja: Abgent
AP11485C
ABGEAP11485CEA 354 EUR
ABGEAP11485C
Anti-TTLL13 Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: TTLL13 (tubulin tyrosine ligase-like family, member 13)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR