Anti-IRGM Rabbit Polyclonal Antibody

Toimittaja: Abgent
AP11128B
ABGEAP11128BEA 354 EUR
ABGEAP11128B
Anti-IRGM Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: IRGM (immunity-related GTPase family, M)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR