Anti-IGHG1 Rabbit Polyclonal Antibody

Toimittaja: Abgent
AP10959C
ABGEAP10959CEA 354 EUR
ABGEAP10959C
Anti-IGHG1 Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: IGHG1 (immunoglobulin heavy constant gamma 1 (G1m marker))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR