Anti-HOXA10 Rabbit Polyclonal Antibody
Toimittaja: Abgent
AP10276C
ABGEAP10276CEA 354 EUR
ABGEAP10276C
Anti-HOXA10 Rabbit Polyclonal Antibody
Vasta-aineet
Type: Primary
Antigen: HOXA10 (homeobox A10)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human