Anti-YY1 Mouse Monoclonal Antibody [clone: 1183CT6.5.23.6]

Toimittaja: Abgent
AM2231B
ABGEAM2231BEA 354 EUR
ABGEAM2231B
Anti-YY1 Mouse Monoclonal Antibody [clone: 1183CT6.5.23.6]
Vasta-aineet
Type: Primary
Antigen: YY1
Clonality: Monoclonal
Clone: 1183CT6.5.23.6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR