Anti-CD3E Mouse Monoclonal Antibody [clone: OKT3]

Toimittaja: Abgent
AM2153B
ABGEAM2153BEA 354 EUR
ABGEAM2153B
Anti-CD3E Mouse Monoclonal Antibody [clone: OKT3]
Vasta-aineet
Type: Primary
Antigen: CD3E
Clonality: Monoclonal
Clone: OKT3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR