Anti-DPPA2 Mouse Monoclonal Antibody [clone: 210CT5.14.6.1]

Toimittaja: Abgent
AM1915B
ABGEAM1915BEA 354 EUR
ABGEAM1915B
Anti-DPPA2 Mouse Monoclonal Antibody [clone: 210CT5.14.6.1]
Vasta-aineet
Type: Primary
Antigen: DPPA2
Clonality: Monoclonal
Clone: 210CT5.14.6.1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR