Anti-THY1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: MRC OX-7]

Toimittaja: Abcam
AB33694-50
ABCAAB33694-50EA 524 EUR
ABCAAB33694-50
Anti-THY1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: MRC OX-7]
Vasta-aineet
Type: Primary
Antigen: THY1 (Thy-1 cell surface antigen)
Clonality: monoclonal
Clone: MRC OX-7
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR