Anti-VE Cadherin Rabbit Polyclonal Antibody
Toimittaja: Abcam
AB33168-100
ABCAAB33168-100EA 475 EUR
ABCAAB33168-100
Anti-VE Cadherin Rabbit Polyclonal Antibody
Vasta-aineet
Rabbit polyclonal to VE Cadherin - Intercellular Junction Marker.
Varoitus: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae,More Product Information
Order Now
TEKNISET TIEDOT
Antigeenin symboli CDH5
Antigeenin nimi cadherin 5, type 2 (vascular endothelium)
Environmentally Preferable
Vasta-aineen tyyppi Ensisijainen
Klonaalisuus Polyclonal
Konjugaatio Unconjugated
Reaktiivisuus Chicken,Human,Mouse
Isäntä Rabbit
Isotyyppi IgG
Western blot Yes
ELISA Yes
ImmunoKemia Yes
ImmunoFluoresenssi Yes
Virtaussytometria Yes