Protein A Agarose (Endotoxin-free)
Toimittaja: Abcam
AB286896-25
ABCAAB286896-25EA 3690 EUR
ABCAAB286896-25
Protein A Agarose (Endotoxin-free)
Elektroforeesigeelit Agaroosi ja akryyliamidi
Reusable, Endotoxin free Protein A conjugated Agarose beads with ≥15 mg/ml binding capacity.
  • APPLICATIONS
  • Purification of monoclonal and polyclonal antibodies from culture media, serum, ascites fluid or hybridoma supernatants
  • Isolation of antibody/antigen complexes in immunoprecipitation experiments, since only the Fc region is involved in antibody binding and the Fab region is available for binding antigen
Endo toxin free Protein A-Agarose beads are prepared by covalently coupling recombinant Protein A to 6% cross-linked Agarose beads underMore Product Information