Anti-THY1 Rabbit Monoclonal Antibody [clone: EPR3133]

Toimittaja: Abcam
AB133350-100
ABCAAB133350-100EA 1180 EUR
ABCAAB133350-100
Anti-THY1 Rabbit Monoclonal Antibody [clone: EPR3133]
Vasta-aineet
Type: Primary
Antigen: THY1 (Thy-1 cell surface antigen)
Clonality: monoclonal
Clone: EPR3133
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR