Anti-IgA Sheep Polyclonal Antibody

Toimittaja: Abcam
AB112881-1
ABCAAB112881-1EA 288 EUR
ABCAAB112881-1
Anti-IgA Sheep Polyclonal Antibody
Vasta-aineet
Type: Secondary
Antigen: IgA (Horse) (horse IgA)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR