Cy7 tetrazine
Toimittaja: AAT BIOQUEST
905
AATB905EA 309 EUR
AATB905
Cy7 tetrazine
Cy7 tetrazine
Cy7 tetrazine is an excellent building block that can be readily used to label TCO-modified biological molecules (e.g., peptides, proteins and oligos etc) for NIR fluorescence in vivo imaging and other fluorescence-based biological applications.
  • Stable for 12 months
Quality tested in-house
Varoitus: For Research Use Only