Fluorescein-dT CPG
Toimittaja: AAT BIOQUEST
6205 6206
AATB6205EA 138 EUR
AATB6205 AATB6206
Fluorescein-dT CPG
Fluorescein-dT CPG
Fluorescein CPG has traditionally been used to add the fluorescein label at the 3'-terminus.
  • Stable for 12 months
Quality tested in-house