ReadiPrep™ PEG Virus Precipitation Kit
Toimittaja: AAT BIOQUEST
60202 60203
AATB60202EA 402 EUR
AATB60202 AATB60203
ReadiPrep™ PEG Virus Precipitation Kit
Bio-assayt Solujen testausanalyysit
Designed for consistent and dependable results. The ReadiPrep™ PEG virus precipitation kit provides a streamlined approach for the concentration of viral particles and the elimination of contaminants, circumventing the need for ultracentrifugation.
  • The use of non-toxic reagents in the procedure ensures safe handling
  • Can concentrate a diverse array of viruses, including retroviruses, baculoviruses, lentiviruses, and phages, from cell culture media and environmental samples, with a concentration increase of over 100-fold
  • Six-month shelf life
This kit is well-suited for both small-scale laboratory samples with low viral titers and large-scale virus preparations with high yields and titers.
Toimitustiedot: Shipped with blue ice.
Varoitus: Research use only.