Buccutite™ Rapid PE-Texas Red Tandem Antibody Labelling Kit
Toimittaja: AAT BIOQUEST
5412
AATB5412EA 2390 EUR
AATB5412
Buccutite™ Rapid PE-Texas Red Tandem Antibody Labelling Kit
Proteiinien yhdistämis- ja leimausjärjestelmät
Production scale optimised for labelling 1 mg antibody per reaction. Buccutite™ Rapid PE-Texas Red tandem antibody labelling kits, designed for large-scale production, provide a streamlined approach for labelling antibodies with PE, APC, PerCP and iFluor® tandem dyes.
  • Robust protocol
  • Consistent results
  • Six-month shelf life
Designed to provide high quality material for downstream fluorescent applications, including flow cytometry.
Toimitustiedot: Shipped with blue ice.
Varoitus: Research use only.