mFluor™ Violet 610 styramide
Toimittaja: AAT BIOQUEST
45079
AATB45079EA 420 EUR
AATB45079
mFluor™ Violet 610 styramide
mFluor™ Violet 610 styramide
mFluor™ Violet 610 Styramide uses the bright red fluorescent dye mFluor™ Violet 610 (Ex/Em = 421/612 nm) to label targets in situ.
  • Stable for 12 months
The Power Styramide™ Signal Amplification (PSA™) system is a highly sensitive method for detecting low-abundant targets in fluorescent immunocytochemistry (ICC), immunohistochemistry (IHC), and in situ hybridization (ISH).
Varoitus: For Research Use Only