5(6)-TAMRA ≥95%

Toimittaja: AAT BIOQUEST
Varoitus

Synonyms: TAMRA 5 and 6-isomer, Tetramethylrhodamine-5-(and-6)-maleimide, 5(6)-Carboxy-tetramethylrhodamine, 9-[2,4(or 2,5)-Dicarboxyphenyl]-3,6-bis(dimethylamino)-xanthylium inner salt

360 361 362
AATB360EA 152 EUR
AATB360 AATB361 AATB362
5(6)-TAMRA ≥95%
5(6)-TAMRA

TAMRA is one of the most popular fluorophores used in various bioconjugations.

Kaava: C₂₅H₂₂N₂O₅
MW: 430.45 g/mol
MDL numero: MFCD16661194
CAS-numero: 98181-63-6
ADR: 5 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1a59cacafc8cb51210444c76e4e24af3b67c0c4c&vid=aa59b976796fe44f82fd97dd98afd367c3601b6e&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR