6-TAMRA ethylenediamine
Toimittaja: AAT BIOQUEST
359
AATB359EA 307 EUR
AATB359
6-TAMRA ethylenediamine
6-TAMRA ethylenediamine
6-TAMRA ethylenediamine is a building block for developing TAMRA bioconjugates.
  • Multiple Bioconjugation Applications
  • Bright Fluorescence
Varoitus: For research use only.