Amplite® West ECL HRP Substrate *Femto Sensitivity*
Toimittaja: AAT BIOQUEST
26100 26101
AATB26100EA 136 EUR
AATB26100 AATB26101
Amplite® West ECL HRP Substrate *Femto Sensitivity*
Elektroforeesireagenssit Western Blotting Reagents
Amplite® West ECL HRP substrate is an ultra-sensitive chemiluminescent (ECL) substrate formulated for highest sensitivity and stability.
  • Stable for 12 months
Quality tested in-house.
Varoitus: For research use only.