Cell Meter™ Autophagy Assay Kit *Green Fluorescence*
Toimittaja: AAT BIOQUEST
23002
AATB23002EA 460 EUR
AATB23002
Cell Meter™ Autophagy Assay Kit *Green Fluorescence*
Bio-assayt Solujen testausanalyysit
This Cell Meter™ autophagy kit employs autophagy Green™ as a specific autophagosome marker to analyse the activity of autophagy.
  • Contains all necessary components
  • Multiple research applications
  • Bright and stable signal
  • Suitable for all major fluorescence platforms
Varoitus: For research use only.