ReadiUse™ Annexin Binding Buffer, Optimised for Imaging and Flow Cytometry
Toimittaja: AAT BIOQUEST
22870 22871
AATB22870EA 111 EUR
AATB22870 AATB22871
ReadiUse™ Annexin Binding Buffer, Optimised for Imaging and Flow Cytometry
Biokemialliset puskuriliuokset Assay Buffers
The ReadiUse™ Annexin Binding Buffer *Optimised for Imaging and Flow Cytometry* is an isotonic buffer designed to promote the labeling of apoptotic cells with annexin V conjugates.
  • Provided as a 10X concentrate
  • Saves time
  • pH of 7.4
Prior to annexin V staining, we ecommend diluting the 10X concentrate to a 1X working solution using distilled water.
Varoitus: For research use only.