MitoLite™ Green EX488
Toimittaja: AAT BIOQUEST
22675
AATB22675EA 152 EUR
AATB22675
MitoLite™ Green EX488
Bio-assayt Solujen testausanalyysit
MitoLite™ Green is a cationic dye that selectively accumulates in mitochondria probably vial the mitochondrial membrane potential gradient.
  • Mitochondria-Specific Dye
  • Cell-Permeable
  • Robust
Varoitus: For research use only.