LysoBrite™ Red DND-99
Toimittaja: AAT BIOQUEST
22647
AATB22647EA 230 EUR
AATB22647
LysoBrite™ Red DND-99
Bio-assayt Solujen testausanalyysit
LysoBrite red DND-99 is chemically same to the LysoTracker® Red DND-99 used for labelling and tracking acidic organelles in live cells (LysoTracker® is the trademark of ThermoFisher).
  • Lysosome-Specific Dye
  • Long-Term Use
  • Low Cytotoxicity
  • Stable After Fixation
Varoitus: For research use only.