Cell Meter™ RX700 Fixable Viability Dye
Toimittaja: AAT BIOQUEST
22532
AATB22532EA 292 EUR
AATB22532
Cell Meter™ RX700 Fixable Viability Dye
Bio-assayt Solujen testausanalyysit
Discrimination and exclusion of dead cells from live cells allows cleaner separation and identification of cell populations.
  • Cell-impermeable
  • Amine- and thiol-reactive
  • High stability
  • Live/dead cell labeling dye
  • Fixable
Varoitus: For research use only.