Cal-520FF™ potassium salt
Toimittaja: AAT BIOQUEST
Varoitus
21144
AATB21144EA 537 EUR
AATB21144
Cal-520FF™ potassium salt
Cal-520FF™ potassium salt
Cal-520® provides a robust homogeneous fluorescence-based assay tool for detecting intracellular calcium mobilization.
  • Improved background/signal ratio
  • Suitable for all major fluorescence platforms
  • Cell impermeant
Varoitus: For research use only.
ADR: 5,II