Fluo-8 AM

Toimittaja: AAT BIOQUEST
Varoitus

21080 21081 21082 21083
AATB21080EA 273 EUR
AATB21080 AATB21081 AATB21082 AATB21083
Fluo-8 AM
Fluo-8 AM

Calcium measurements are critical for numerous biological investigations.


  • Can be loaded at rt
  • Improved calcium response
  • Significantly increased signal
  • Suitable for all fluorescence platforms


Varoitus: For research use only.

CAS-numero: 1345980-40-6
ADR: 5,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR