ReadiUse™ Microscope Mounting Solution (50 ml)
Toimittaja: AAT BIOQUEST
20009
AATB20009EA 120 EUR
AATB20009
ReadiUse™ Microscope Mounting Solution (50 ml)
Värit Microscopy and staining reagents
The mounting medium is the solution in which the specimen is embedded, generally under a cover glass.
  • Non-reactive
  • Long-term stability
  • Minimal hands-on time
  • Ready to use
Varoitus: For research use only.