EDTA, AM ester metal ion chelator

Toimittaja: AAT BIOQUEST
Varoitus

19010 19011
AATB19010EA 152 EUR
AATB19010 AATB19011
EDTA, AM ester metal ion chelator
EDTA, AM ester

Ethylenediaminetetraacetic acid (EDTA) might be the most popular metal ion chelator used for both industrial and medical purposes.


  • Excellent metal chelator
  • Multiple applications

CAS-numero: 162303-59-5
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0ec4f685e9f6d8fcd7cae9a30a2126881928e46f&vid=6155ffacf55e8a1673beb43ba642717ddca623e5&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR