dGTP *100 mM PCR Grade*
Toimittaja: AAT BIOQUEST
17254 17255
AATB17254EA 138 EUR
AATB17254 AATB17255
dGTP *100 mM PCR Grade*
Nukleiinihapporeagenssit Nukleotidit
2'-Deoxyguanosine-5'-Triphosphate (dGTP) solution is used in DNA synthesis reactions such as PCR, DNA sequencing and molecular cloning techniques.
  • Highly purified
DNA sequencing is a fundamental tool in biological and medical research (especially for personalised medicine).
Varoitus: For research use only.