dCTP *100 mM PCR Grade*
Toimittaja: AAT BIOQUEST
17252 17253
AATB17252EA 138 EUR
AATB17252 AATB17253
dCTP *100 mM PCR Grade*
Nukleiinihapporeagenssit Nukleotidit
2'-Deoxycytidine-5'-Triphosphate (dCTP) solution is used in DNA synthesis reactions such as PCR, DNA sequencing and molecular cloning techniques.
  • Highly purified
DNA sequencing is a fundamental tool in biological and medical research (especially for personalised medicine).
Varoitus: For research use only.