Fluorescein-12-dUTP (Perkin-Elmer) *1 mM in TE Buffer (pH 7,5)*
Toimittaja: AAT BIOQUEST
17027-100NM 17027-1UM
AATB17027-100NMEA 458 EUR
AATB17027-100NM AATB17027-1UM
Fluorescein-12-dUTP (Perkin-Elmer) *1 mM in TE Buffer (pH 7,5)*
Nukleiinihapporeagenssit Nukleotidit
This fluorescein-12-dUTP conjugate can be used as a green fluorescence color with Spectrum Green™ filter set (Spectrum Green™ is the trademark of Vysis).
  • Chemically identical to fluorescein-12-dUTP (Perkin Elmer’s NEL413001EA)
The dye-modified deoxyuridine 5'-triphosphates are one of the most common methods to produce dye-labelled DNA via conventional enzymatic incorporation methods such as reverse transcription, nick translation, random primed labelling, or PCR.
Varoitus: For research use only.