mFluor™ Violet 450 azide
Toimittaja: AAT BIOQUEST
1690
AATB1690EA 429 EUR
AATB1690
mFluor™ Violet 450 azide
mFluor™ Violet 450 azide
mFluor™ Violet 450 dyes have fluorescence excitation and emission maxima of ~405 nm and ~450 nm respectively.
  • Stable for 12 months
AAT Bioquest's mFluor™ dyes are developed for multicolor flow cytometry-focused applications.
Varoitus: For Research Use Only