SunRed™ Phosphate
Toimittaja: AAT BIOQUEST
Varoitus
11629
AATB11629EA 307 EUR
AATB11629
SunRed™ Phosphate
SunRed™ Phosphate
Phosphatase-catalyzed hydrolysis of Sun Red phosphate (SRP) yields the Sun Red fluorophore that can be excited with the 633 nm laser with emission of ~660 nm.
  • Optimised for Cy5 filter set
  • Extremely sensitive NIR esterase substrate
  • Excellent cell viability stain
  • Useful in combination with green vital stains
ADR: 5,II