Anti-CD161 Mouse Monoclonal Antibody [clone: HP-3G10] (PB (Pacific Blue))

Toimittaja: AAT BIOQUEST
116101J0 116101J1
AATB116101J0EA 371 EUR
AATB116101J0 AATB116101J1
Anti-CD161 Mouse Monoclonal Antibody [clone: HP-3G10] (PB (Pacific Blue))
Vasta-aineet
HP-3G10 is an anti-human monoclonal antibody that recognizes the CD161 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Varoitus: For research use only.

Type: Primary
Antigen: CD161
Clonality: Monoclonal
Clone: HP-3G10
Conjugation: PB (Pacific Blue)
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR