Fluorescein diphosphate tetraammonium salt (FDP) ≥95%

Toimittaja: AAT Bioquest
Varoitus

11600 11601
AATB11600EA 564 EUR
AATB11600 AATB11601
Fluorescein diphosphate tetraammonium salt (FDP) ≥95%
Fluorescein diphosphate tetraammonium salt (FDP)

Upon interaction with phosphatases the colourless and non-fluorescent FDP is hydrolysed to highly fluorescent fluorescein, which exhibits excellent spectral properties that match the optimal detection window of most fluorescence instruments that are equipped with the Argon laser excitation.


  • Useful for multiple applications

Kaava: C₂₀H₂₆N₄O₁₁P₂
Storage Temperature: Pakastin
MDL numero: MFCD00278758
CAS-numero: 217305-49-2
ADR: 5,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR